Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermo Scientific™ Bioreactor Tubes, 50 ml

Supplier: Thermo Fisher Scientific
Thermo Scientific™
332260
77392-120CS 121.7 USD InStock
77392-120
Thermo Scientific™ Bioreactor Tubes, 50 ml
Tubes Culture Tubes
Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition.

  • Polypropylene tube
  • HDPE cap
  • 2 µm PTFE filter membrane
  • Polystyrene dial
  • Suitable for suspension culture
  • Sterile - SAL 10⁻⁶
  • Non-pyrogenic
  • Non-cytotoxic
  • Shelf life two years

Ideal for screening, small volume high throughput analysis. Protein expression suspension culture volumes ranging from 10 to 25 ml. Convenient pack sizes. Four different sizes of ventilation (dial type) on the cap.

Packaging: 3 per pack; 30 packs per case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty