Would you like to log in again?
VWR® Clinical Centrifuge for Blood Banking, Serology 12
Supplier: Avantor
VWR®
00-483-037-001
77392-164EA 4569.11 USD InStock
77392-164
VWR® Clinical Centrifuge for Blood Banking, Serology 12
Centrifuges Benchtop Centrifuges
The only drop-in replacement for a best selling blood banking centrifuge. Perfect for all blood typing and cross-matching activities, the serology 12 integrates into any laboratory with minimal process changes.
  • View time remaining and set cycle parameters using the digital display
  • Store up to 10 custom time, speed, acceleration and braking settings
  • Max speed 1040×g/3550 rpm
  • Spin up to 6 capped and 12 uncapped 13×75 mm tubes with minimal handling
  • Tube positions are numbered for quick identification
  • Compatible with existing SeroFuge rotors
  • 2 year warranty
The VWR® serology 12 clinical centrifuge is specifically designed to facilitate high-quality blood grouping, typing, cross matching and other cell-washing procedures for a wide range of tube sizes. Based on a popular blood banking centrifuge and using the same rotors, the serology 12 ensures samples are efficiently processed, with no individual handling necessary.
Certifications: CE, MET, UKCA.
Delivery information: Supplied with 12 tube rotor and tube holders.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty