Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SMALP BZ Screening Kit

Supplier: Cube Biotech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

16690
77392-442EA 416 USD InStock
77392-442
SMALP BZ Screening Kit
Protein Expression Protein Expression Reagents
SMALP BZs are synthesized via RAFT polymerization, achieving a better-defined polymer size and a low dispersity.

  • Usage: Membrane protein solubilization and stabilization
  • Full-Name: Poly(styrene-co-maleic acid-co-X%(N-benzyl)maleimide) copolymer, sodium salt in 50 mM HEPES, pH 7.5; X is be replaced with the number in the products name
  • Formula Weight: 5500 g/mol
  • Buffer: 50 mM HEPES
  • pH: 7.5
  • State: Lyophilized powder
  • Recommended concentration range upon use: 1 to 5%
  • Solubility: >10% in H₂O
  • Color: White to slightly yellow
  • Odor: Odorless

The versatility of membrane proteins and their composition mirrors the intricate nature of the cell membrane. As a result, it is a challenge to predict with absolute certainty the optimal SMALP BZ polymer for the solubilization and subsequent stabilization of your membrane protein of interest. Therefore, we offer the SMALP BZ Screening Kit that consists of our  four individual SMALP BZs. This way you can conveniently screen for the best SMALP BZ for your protein before scaling up your assay. The SMALP BZ series was developed in collaboration with Prof. Bert Klumperman from Nanosene, a spinout company of Stellenbosch University in South Africa.

Delivery information: Shipping temperature: Ambient temperature; Storage of lyophilized copolymer: –20 °C for several years; Storage of dissolved copolymer: 2 to 8 °C for several day.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty