Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HiYield Cell-free E. coli Lysate

Supplier: Cube Biotech
HiYield
21013 21011
77392-648PK 648 USD InStock
77392-648 77392-646
HiYield Cell-free E. coli Lysate
Protein Purification Protein Purification Reagents
Our E. coli cell-free lysates are the isolated protein purification machinery of bacterial cells. They create a high-yielding and adjustable cell-free protein expression system.

  • Origin of the MSP protein: Mouse
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 388
  • Molecular mass: 45526 Da
  • Extinction coefficient (in water) ε280: 39880 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Affinity tag: N-Terminal His-tag

The advantages of cell-free protein expression are numerous, among other things it enables a scientist to:
- For direct stabilization and co-translational integration of membrane proteins into nanodiscs
- Expression of otherwise toxic proteins

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty