Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nanodisc Assembly Kit MSP1D1 dH5 POPC

Supplier: Cube Biotech
26243
77392-700PK 755 USD InStock
77392-700
Nanodisc Assembly Kit MSP1D1 dH5 POPC
Protein Expression Protein Expression Reagents
This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP1D1 dh5 and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.

  • Origin of MSP protein: Human
  • Purity: >90% (determined by SDS-PAGE)
  • Number of amino acids: 392
  • Molecular mass: 45670 Da
  • Extinction coefficient (in water) ε280: 44920 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (human origin): GHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • Affinity tag: N-Terminal His-tag

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dh5 creates nanodiscs of ∼7 to 8 nm in diameter.

With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs.

Delivery information: Shipping temperature: Room temperature; storage of lyophilized copolymer: −20 °C for several years; storage of reconstituted protein: 2 - 8 °C for several day.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty