Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nanodisc MSP2N2 DMPC

Supplier: Cube Biotech
26381
77392-752EA 995 USD InStock
77392-752
Nanodisc MSP2N2 DMPC
Protein Expression Protein Expression Reagents
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.

  • Origin of the MSP protein: Human
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 371
  • Molecular mass: 43.05 kDa
  • Extinction coefficient (in water) ε280: 36900 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (1 Letter code): G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
  • Affinity tag: none

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 creates nanodiscs of ∼17 nm in diameter.

Delivery information: Shipping temperature: Dry-ice; Storage of reconstituted protein: –80 °C, avoid freeze-thaw cycles.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty