Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nanodisc Rat MSP1E3D1-His DMPG

Supplier: Cube Biotech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

26955
77392-830EA 768 USD InStock
77392-830
Nanodisc Rat MSP1E3D1-His DMPG
Protein Expression Protein Expression Reagents
Membrane scaffold protein. Rat origin.

  • Origin of the MSP protein: Rat
  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 267
  • Molecular Mass: 31572.76 Da
  • Extinction coefficient (in water) ε280: 36440 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Affinity tag: N-Terminal His-tag

Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (rat) creates nanodiscs of ∼12 to 14 nm in diameter. This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein. There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.

Delivery information: Shipping Temperature: Dry-Ice. Storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty