Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Ni-IDA MagBeads

Supplier: Cube Biotech
PureCube
30290 30291 30225 30205 30201
77392-904EA 72 USD InStock
77392-904 77392-906 77392-912 77392-910 77392-908
PureCube Ni-IDA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Ni-IDA MagBeads were developed by Cube Biotech for His-tag protein purification.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead ligand: anti-Rho1D4 antibody
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C

Our year-long experience manufacturing agarose resin led to a high yield of 70 mg protein per ml resin, leading in the market compared to other Ni-IDA suppliers. Ni-IDA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin or as XL magnetic beads for more viscous cell media. For convenience, Cube Biotech also offers its own MagBead separator for use with our magnetic beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty