Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Ni-IDA

Supplier: Cube Biotech
PureCube
30304 30308 30302 30306
77392-916PK 241 USD InStock
77392-916 77392-918 77392-914 77392-920
PureCube Compact Cartridge Ni-IDA
Protein Purification Protein Purification Reagents
Our PureCube Ni-IDA XL agarose resins are agarose resin beads with a diameter of ∼400 µm. They purify active His-tagged proteins from cells and extremely viscous cell media.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >70 mg/ml
  • Bead Ligand: Ni-IDA
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
  • Required equipment and materials:
  • FPLC hardware like e.g. ÄKTA
  • Binding Buffer
  • Elution Buffer

Our year-long experience manufacturing agarose resins led to a high yield of 20 mg protein per ml resin, leading in the market compared to other Ni-IDA suppliers. PureCube Ni-IDA resins are suited for batch spin columns and FPLC. Their small diameter provides them with excellent mechanical stability. We recommend our Ni-IDA agarose beads with a 40 µm diameter for standard-sized beads with higher protein yield. Furthermore, it is also possible to get the 40 µm Ni-IDA beads pre-packed in a column/cartridge or as magnetic beads. The PureCube Cartridge is compatible with standard FPLC systems like ÄKTA.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty