Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube IDA MagBeads

Supplier: Cube Biotech
PureCube
30805 30890 30801 30825
77392-938EA 1040 USD InStock
77392-938 77392-934 77392-936 77392-940
PureCube IDA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube IDA (iminodiacetic acid) MagBeads were developed for IMAC-based protein purification and protein enrichment methods.

  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: IDA
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

The beads in this suspension have an average diameter of 30 µm and, thus, excellent mechanical stability. Since the affinity of an IDA-metal-ion-complex depends on the loaded metal ion, we offer IDA MagBeads uncoupled to any metal ion for the case that we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach desired metal ions to the agarose beads. PureCube IDA MagBeads are also available with a bead diameter of 90 µm. PureCube IDA products are also obtainable as agarose resin for FPLC assays or batch spin approaches.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty