Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Co-NTA MagBeads

Supplier: Cube Biotech
PureCube
31525 31505 31590 31501
77392-990EA 113 USD InStock
77392-990 77392-996 77392-992 77392-994
PureCube Co-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Co-NTA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.

  • Usage: Specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead Ligand: Co-NTA
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Our year-long experience in manufacturing agarose resin led to the high yield of 30 mg protein per ml resin, leading in the market compared to other Co-NTA suppliers. Co-NTA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin or as XL magnetic beads for more viscous cell media. For convenience, Cube Biotech also offers its own MagBead separator for use with our magnetic beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty