Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Co-NTA

Supplier: Cube Biotech
PureCube
31608 31602 31606 31604
77393-002EA 187 USD InStock
77393-002 77393-000 77392-998 77393-004
PureCube Compact Cartridge Co-NTA
Protein Purification Protein Purification Reagents
The PureCube Co-NTA Compact cartridge is an IMAC chromatography column prepacked with PureCube Co-NTA resin.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead Ligand: Co-NTA
  • Bead size: 40 μm
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Filling quantity: Delivered as a 50% suspension
  • Required equipment and materials:
  • FPLC hardware like e.g. ÄKTA
  • Binding Buffer
  • Elution Buffer

You can choose between two sizes:1 ml bed volume and 5 ml bed volume. The PureCube cartridge/column used here is compatible with almost all commonly used FPLC systems like ÄKTA. If not, we recommend buying the sole loose PureCube Co-NTA resin and packing the columns yourself. Alternatively, we offer our PureCube Co-NTA products as magnetic beads if you prefer this IMAC method.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty