Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Glutathione MagBeads

Supplier: Cube Biotech
PureCube
32201 32205 32290 32225
77393-050EA 1244 USD InStock
77393-050 77393-052 77393-046 77393-048
PureCube Glutathione MagBeads
Protein Purification Protein Purification Magnetic Beads
The protein purification with the PureCube Glutathione MagBeads and the GST-tag exploit the specific enzyme-substrate binding to gain high levels of purity while maintaining a high protein yield.

  • Usage: Specific binding and purification of GST-tagged proteins
  • Specificity: Affinity to GST(Glutathione-S-Transferase)-tagged proteins
  • Binding capacity: >20 mg/ml
  • Bead Ligand: L-Glutathione (reduced)
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH Stability: 3 to 12
  • Stabilities: 1 M sodium acetate, pH 4.0, or 6 M guanidinehydrochloride, organic solvents (e.g., 70% (v/v) ethanol), 1% (w/v) SDS, 0.1 M NaOH, 0.1 M HCl

The pairing of an enzyme to its substrate is one of the most specific combinations in life sciences. PureCube Glutathione MagBeads is a suspension of tiny 30 µm magnetic purification beads that are used for this purpose. Their small size provides high mechanical stability. Additionally, we offer GST-tag affinity MagBeads with a bead diameter of 90 µm for very viscous cell media. Alternatively, Glutathione Agarose resins for FPLC or batch spin approaches are also available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty