Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Glutathione

Supplier: Cube Biotech
PureCube
32304 32306 32302 32308
77393-060PK 1163 USD InStock
77393-060 77393-054 77393-058 77393-056
PureCube Compact Cartridge Glutathione
Protein Purification Protein Purification Reagents
The protein purification with the PureCube Glutathione agarose and the GST-tag exploit the specific enzyme-substrate binding to gain high levels of purity while maintaining a high protein yield.

  • Usage: Specific binding and purification of GST-tagged proteins
  • Specificity: Affinity to GST(Glutathione-S-Transferase)-tagged proteins
  • Binding capacity: >10 mg/ml
  • Bead Ligand: L-Glutathione (reduced)
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • pH Stability: 3 to 12
  • Required equipment and materials:
  • FPLC hardware like e.g. ÄKTA
  • Binding Buffer
  • Elution Buffer

The pairing of an enzyme to its substrate is one of the most specific combinations in life sciences. The PureCube cartridge/column used here is compatible with almost all commonly used FPLC systems like ÄKTA. The PureCube Glutathione Compact cartridge is an FPLC column pre-packed with PureCube Glutathione Agarose. Alternatively, Glutathione MagBeads are also available if you prefer this affinity chromatography approach. Additional support products for GST-tag purification assays are: L-glutathione (reduced), essential compound of the elution buffer, GST-tag antibodies for western blot detection.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty