Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HighSpec Rho1D4 Agarose

Supplier: Cube Biotech
HighSpec
33102 33101 33104 33103
77393-064EA 1241 USD InStock
77393-064 77393-062 77393-068 77393-066
HighSpec Rho1D4 Agarose
Protein Purification Protein Purification Reagents
HighSpec Rho1D4 Agarose resin is a suspension of 40 µm agarose beads, coupled to an anti-Rho1D4 antibody. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead Ligand: anti-Rho1D4 antibody
  • Bead size: 100 μm
  • Matrix: 6% cross-linked agarose
  • Recommended flow rates: 0.25 to 1 ml/min
  • Max. flow rates: 6 ml/min
  • Filling quantity: Delivered as a 50% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C
  • Required equipment:
  • Lysis Buffer
  • Wash Buffer
  • Elution Buffer (incl. Rho1D4 peptide)
  • Ice bath
  • Refrigerated centrifuge for 50 ml tube (min 10000×g)
  • 50 ml centrifuge tube
  • Micropipettor and Micropipetting tips
  • Disposable gravity flow columns with capped bottom outlet, 2 ml
  • pH meter
  • End-over-end shaker
  • SDS-PAGE buffers, reagents and equipment Optional: Western Blot reagents and equipment

Protein affinity epitope tags find use in numerous applications in life sciences. Ranging from Western Blot detection, protein localization, ELISA assays, and most importantly protein purification. The possible uses for epitope tags are numerous (Molday and Molday 2014). Rho1D4, sometimes also just called '1D4-tag', is an epitope tag that originates from the C-terminal end of bovine rhodopsin. Its nine amino acids long sequence is TETSQVAPA, which gives it a size of 902.9 Dalton. The affinity tag and the specific monoclonal antibodies were discovered in 1984 by Hodges et al. and MacKenzie et al.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty