Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HighSpec HighSpec Rho1D4 MagBeads

Supplier: Cube Biotech
HighSpec
33225 33205 33201
77393-072EA 404 USD InStock
77393-072 77393-076 77393-074
HighSpec HighSpec Rho1D4 MagBeads
Protein Purification Protein Purification Magnetic Beads
HighSpec Rho1D4 MagBeads are a suspension of 30 µm magnetic agarose beads coupled to an anti-Rho1D4 antibody.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead Ligand: Anti-Rho1D4 antibody
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Recommended flow rates: 0.25 to 1 ml/min
  • Max. flow rates: 6 ml/min
  • Filling quantity: Delivered as a 50% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C

Required equipment and materials:
- FPLC hardware like e.g. AKTA
- Binding buffer
- Elution buffer

HighSpec Rho1D4 MagBeads are meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. Additionally, we offer Rho1D4-tag affinity resin with a bead diameter of 90 µm for very viscous cell media. Alternatively, Rho1D4 Agarose resin is also available if you prefer this affinity chromatography method.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty