Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HighSpec Rho Starter Set 3: HighSpec Compact Cartridge Rho1D4 + Rho1D4 Peptide

Supplier: Cube Biotech
HighSpec
33398
77393-088PK 320 USD InStock
77393-088
HighSpec Rho Starter Set 3: HighSpec Compact Cartridge Rho1D4 + Rho1D4 Peptide
Protein Purification Protein Purification Reagents
Our Rho1D4 starter set 3 combines our HighSpec Rho1D4 compact cartridge and the Rho1D4 elution peptide.

  • Usage: As a matrix for Hydrophobic Interaction Chromatography (HIC)
  • Specificity: Hydrophobic biomolecules and proteins
  • Bead size: 40 μm
  • Bead ligand: Butyl groups
  • Butyl group density: 40 µmol/ml
  • BSA binding capacity: 25 mg/ml
  • Lysozyme binding capacity: 45 mg/ml
  • pH stability: 2 to 13
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Chemical structure of butyl ligand: Butyl group structure

The compact cartridge is an FPLC column pre-packed with our HighSpec Rho1D4 agarose resin. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty