Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube HiCap StrepTactin® MagBeads

Supplier: Cube Biotech
PureCube StrepTactin®
34201 34205 34225 34290
77393-098EA 562 USD InStock
77393-098 77393-096 77393-102 77393-100
PureCube HiCap StrepTactin® MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube StrepTactin® MagBeads are a suspension of 6% cross-linked magnetic agarose beads coupled to a StrepTactin protein.

  • Usage: Highly specific affinity purification of Strep-(WSHPQFEK)-tagged proteins
  • Specificity: Affinity to Strep-tagged (WSHPQFEK) proteins
  • Binding capacity: >7 mg/ml
  • Bead ligand: StrepTactin®
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 5 % suspension
  • Chemical stability: 2% Tween 20, 2% Triton X-100, 2% IGEPAL® 630/Nonidet P40, n-Octyl-beta-D-glucopyranoside, 0,2% n-Nonyl-beta-D-glucopyranoside, 0,35% N-Decyl-beta-D-maltoside, 2% Lauryl-sarcosine, 0, 1% SDS, 0,3% CHAPS, 1M Guanidine HCl, 1mM PMSF, 10% Ethanol, 5M NaCl, 2M (NH4)SO₄, 1M CaCl₂, 25% Glycerol
  • Short-term storage: In equilibration buffer
  • Long-term storage: In 20% ethanol at 4 °C

PureCube StrepTactin® MagBeads enable protein yields of up to 7 mg protein per ml MagBeads and were developed for protein purification assays using the Strep-tag® and magnetic separation. PureCube StrepTactin® products are also obtainable as agarose resin. Furthermore, PureCube StrepTactin® MagBeads XL are also available for very vicious cell media.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty