Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Phenyl Agarose

Supplier: Cube Biotech
PureCube
39105 39115 39110 39112 39103
77393-130EA 1680 USD InStock
77393-130 77393-124 77393-132 77393-128 77393-126
PureCube Phenyl Agarose
Protein Purification Protein Purification Reagents
Our PureCube phenyl agarose resin is a suspension of purification beads for hydrophobic interaction chromatography.

  • Antibody name: Rho-1d4-1
  • Cell line/origin: Mouse, hybridoma
  • Antibody purification: rProtein A
  • Quantity: 200 µg
  • Concentration: 4.11 mg/ml (by UV absorbance at 280 nm)
  • Physical State: Liquid (sterile filtered)
  • Color: Clear
  • Buffer: 0.25 M sodium bicarbonate, 0.5 M sodium chloride, pH = 8.3
  • Purity: >95% (determined by non-denaturing SDS-PAGE)
  • Shipment temperature: Shipped on ice
  • Short-term storage: Store vial at 4 °C. Centrifuge product if not completely clear after standing at room temperature and/or heating to 37 °C
  • Long-term Storage: Aliquot contents and freeze at –20 °C or below. Avoid repeated freeze-thaw cycles

Hydrophobic Interaction Chromatography (HIC) is a reliable method to separate proteins and other biomolecules based on their hydrophobicity. If you use HIC with your sample for the first time we recommend testing all of our HIC resins before.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty