Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Octyl Agarose

Supplier: Cube Biotech
PureCube
39203 39215 39205 39210 39212
77393-142PK 3103 USD InStock
77393-142 77393-140 77393-134 77393-138 77393-136
PureCube Octyl Agarose
Protein Purification Protein Purification Reagents
Our PureCube octyl agarose resin is a suspension of purification beads for Hydrophobic Interaction Chromatography (HIC).

  • Antibody name: PentaHis antibody, BSA-free
  • Cell Line: Mouse, hybridoma
  • Suitable for: Western blot, ELISA
  • Detection limit Western blot, chromogenic detection: 2 ng
  • Detection limit Western blot, chemiluminescent detection: 50 pg
  • Quantity: 100 µg
  • Physical state: Dried lyophilized powder
  • Reconstitution of lyophilized antibody: Add 500 µl ddH₂O to obtain a solution of 0.2 mg/ml concentration
  • Recommended working dilution: 1:1000
  • Shipment temperature: Shipped at ambient temperature
  • Short-term storage: After resuspension, store vial at 2 to 8 °C for up to 3 months. Centrifuge product if not clear after standing at room temperature and/or heating to 37 °C
  • Long-term storage: Store as lyophilized powder for up to 12 months at 2 to 8 °C. Alternatively, aliquot contents and freeze at –20 °C for up to six months. Avoid repeated freeze-thaws

Hydrophobic Interaction Chromatography (HIC) is a reliable method to separate proteins and other biomolecules based on their hydrophobicity. If you use HIC with your sample for the first time we recommend testing all of our HIC resins before.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty