Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Carboxy MagBeads

Supplier: Cube Biotech
PureCube
50225 50290 50201 50205
77393-170PK 7112 USD InStock
77393-170 77393-164 77393-168 77393-166
PureCube Carboxy MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube amine carboxy agarose MagBeads are your best option to couple biomolecules that carry free-standing Amine (NH₂) groups.

  • Usage: Coupling biomolecules to active Carboxy Groups
  • Specificity: Exposed Amine (NH₂) groups on biomolecule
  • Amine group density: Higher than 15 µmol/ml
  • Bead: Ligand chemically active Carboxy groups
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Matrix: 6% cross-linked agarose
  • Short term storage: In equilibration buffer (see protocol)
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C
  • or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three years upon delivery, unopened
  • Chemical structure: Carboxy structure

The resulting peptide bond between the Carboxy MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Carboxy MagBeads are also available as agarose resin if you prefer this purification method or as XL MagBeads if your downstream application involves a very viscous cell media.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty