Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube NHS Activated Agarose

Supplier: Cube Biotech
PureCube
50310 50305 50303 50315 50312
77393-180PK 10114 USD InStock
77393-180 77393-174 77393-172 77393-178 77393-176
PureCube NHS Activated Agarose
Protein Purification Protein Purification Reagents
PureCube NHS (N-hydroxy succinimide) Agarose resin is your best option to couple biomolecules with free-standing amine groups.

  • Usage: Coupling proteins to active NHS Groups
  • Specificity: Exposed amine (NH₂) groups on biomolecule
  • Binding capacity: >20 mg/ml
  • Bead Ligand: Chemically active NHS groups
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • Storage Buffer: 100% isopropanol
  • Matrix: 7.5% cross-linked agarose
  • Storage Temperature: 4 °C
  • Shelf life: If properly stored - 6 months upon delivery, unopened
  • Chemical Structure: NHS structure
  • Special Notes: This product is shipped as a flammable good, 2-propanol UN1219

The resulting Amide bond between the NHS Agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube NHS Activated Agarose is also available as magnetic beads if you prefer this purification method or as XL agarose resin if you need higher flow rates for your downstream application.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty