Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Amine Activated MagBeads

Supplier: Cube Biotech
PureCube
50905 50925 50990 50901
77393-208EA 690 USD InStock
77393-208 77393-206 77393-212 77393-210
PureCube Amine Activated MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube amine activated MagBeads are your best option to couple biomolecules that carry free-standing carboxy (COOH) groups.

  • Usage: Coupling proteins to active Amine Groups
  • Specificity: Exposed carboxy (COOH) groups on biomolecule
  • Amine group density: Higher than 20 µmol/ml
  • Bead ligand: Chemically active Amine groups
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 6% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three years upon delivery, unopened
  • Chemical structure: Amine structure

The resulting peptide bond between the Amine MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Amine Activated MagBeads are also available as agarose resin if you prefer this purification method or as XL MagBeads if your downstream application involves more viscous cell media.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty