Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Amine Activated Agarose

Supplier: Cube Biotech
PureCube
51012 51005 51010 51003
77393-218EA 2700 USD InStock
77393-218 77393-220 77393-216 77393-214
PureCube Amine Activated Agarose
Protein Purification Protein Purification Reagents
PureCube amine activated agarose resin is your best option to couple biomolecules carrying free-standing carboxy (COOH) groups.

  • Usage: Coupling proteins to active Amine Groups
  • Specificity: Exposed carboxy (COOH) groups on biomolecule
  • Amine group density: Higher than 20 µmol/ml
  • Bead ligand: Chemically active Amine groups
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - 3 years upon delivery, unopened
  • Chemical Structure: Amine structure

The resulting peptide bond between the Amine Agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Amine Activated Agarose is also available as magnetic beads if you prefer this purification method or as XL agarose resin if you need higher flow rates for your downstream application.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty