Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Maleimide Activated Agarose

Supplier: Cube Biotech
PureCube
51103 51112 51105 51115 51110
77393-228EA 14184 USD InStock
77393-228 77393-230 77393-222 77393-226 77393-224
PureCube Maleimide Activated Agarose
Protein Purification Protein Purification Reagents
PureCube Maleimide Agarose resin is your best option to couple biomolecules that carry thiol (SH) groups.

  • Usage: Coupling proteins to active Maleimide Groups
  • Specificity: Exposed thiol (SH) groups on biomolecule
  • Bead Ligand: Chemically active Maleimide groups
  • Resulting Bond: irreversible Thioether
  • Protein binding capacity: ≥15 mg/ml
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% cross-linked agarose
  • Short Term Storage: In neutral buffer at 4 °C
  • Long Term Storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - 3 months upon delivery, unopened
  • Chemical Structure: Maleimide structure

The resulting Thioether bond between the Maleimide Agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Maleimide Activated Agarose is also available as magnetic beads if you prefer this method or as XL agarose resin if you need higher flow rates for your downstream application.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty