Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Ni-IDA MagBeads XL

Supplier: Cube Biotech
PureCube
55205 55290 55225
77393-260PK 4334 USD InStock
77393-260 77393-256 77393-258
PureCube Ni-IDA MagBeads XL
Protein Purification Protein Purification Magnetic Beads
PureCube Ni-IDA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.

  • Usage: Specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity: >40 mg/ml
  • Bead Ligand: Ni-IDA
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Our year-long experience in manufacturing agarose resin lead to the high yield of 40 mg protein per ml MagBeads. The XL beads are meant for viscous media and cell debris.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty