Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube NTA MagBeads XL

Supplier: Cube Biotech
PureCube
55990 55905 55925
77393-300EA 1979 USD InStock
77393-300 77393-298 77393-302
PureCube NTA MagBeads XL
Protein Purification Protein Purification Magnetic Beads
PureCube NTA (nitrilotriacetic acid) XL MagBeads were developed for IMAC-based protein purification and protein enrichment methods.

  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: NTA
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

The beads in this suspension have an average diameter of 90 µm. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA MagBeads uncoupled to any metal ion for the case that we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty