Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Alprenolol Agarose

Supplier: Cube Biotech
PureCube
56031 56032 56034 56035
77393-308EA 6026 USD InStock
77393-308 77393-306 77393-310 77393-304
PureCube Alprenolol Agarose
Protein Purification Protein Purification Reagents
PureCube Alprenolol resin provides highly-specific purification β-adrenergic receptors with maximum protein yield.

  • Usage: Specific binding and purification of β-adrenergic receptors
  • Specificity: Affinity to β-adrenergic receptors
  • Bead ligand: Alprenolol
  • Bead size: 100 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Storage media: Contains 0.02% sodium azide
  • Storage: pH 7.5
  • Long-term storage: 100 mM sodium hydrogencarbonate, 0.02% sodium azide, pH 7.5, ethanol at 4 °C

Protein purification always aims for maximum purity and the highest protein yield. However, often, these two stand against each other. The solution to this problem is to find a specific ligand for your protein that does not require an additional affinity tag to your protein while being small enough that an agarose bead can be covered with this ligand to a great extent. This solution is one of them.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty