Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube HiCap StrepTactin® Agarose XL

Supplier: Cube Biotech
PureCube StrepTactin®
57303 57310 57305
77393-328EA 5168 USD InStock
77393-328 77393-330 77393-326
PureCube HiCap StrepTactin® Agarose XL
Protein Purification Protein Purification Reagents
Our PureCube HiCap StrepTactin® agarose XL resin beads consist of 6% cross-linked agarose coupled to the StrepTactin protein.

  • Usage: Highly specific affinity purification of Strep-(WSHPQFEK)-tagged proteins
  • Specificity: Affinity to Strep-tagged (WSHPQFEK) proteins
  • Binding capacity: >5 mg/ml
  • Bead Ligand: StrepTactin®
  • Bead size: 400 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Chemical stability: 2% Tween 20, 2% Triton X-100, 2% IGEPAL® 630/Nonidet P40, n-Octyl-beta-D-glucopyranoside, 0,2% n-Nonyl-beta-D-glucopyranoside, 0,35% N-Decyl-beta-D-maltoside, 2% Lauryl-sarcosine, 0, 1% SDS, 0,3% CHAPS, 1M Guanidine HCl, 1mM PMSF, 10% Ethanol, 5M NaCl, 2M (NH4)SO4, 1M CaCl2, 25% Glycerol
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with sodium azide at 4 °C
  • Required equipment:
  • Lysis Buffer
  • Wash Buffer
  • Elution Buffer (incl. Rho1D4 peptide)
  • Ice bath
  • Refrigerated centrifuge for 50 ml tube (min 10000×g)
  • 50 ml centrifuge tube
  • Micropipettor and Micropipetting tips
  • Disposable gravity flow columns with capped bottom outlet, 2 ml
  • pH meter
  • End-over-end shaker
  • SDS-PAGE buffers, reagents and equipment Optional: Western Blot reagents and equipment

The PureCube HiCap StrepTactin® agarose XL resin beads enable protein yields of up to 5 mg protein per ml resin and were developed for FPLC and batch spin-based protein purification.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty