Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube HiCap StrepTactin® MagBeads XL

Supplier: Cube Biotech
PureCube StrepTactin®
57405 57490 57425
77393-332EA 676 USD InStock
77393-332 77393-336 77393-334
PureCube HiCap StrepTactin® MagBeads XL
Protein Purification Protein Purification Magnetic Beads
PureCube StrepTactin® MagBeads XL is a suspension of 6% cross-linked magnetic agarose beads coupled to the StrepTactin protein.

  • Usage: Highly specific affinity purification of Strep-(WSHPQFEK)-tagged proteins
  • Specificity: Affinity to Strep-tagged (WSHPQFEK) proteins
  • Binding capacity: >7 mg/ml
  • Bead Ligand: StrepTactin®
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 5% suspension
  • Chemical stability: 2% Tween 20, 2% Triton X-100, 2% IGEPAL® 630/Nonidet P40, n-Octyl-beta-D-glucopyranoside, 0,2% n-Nonyl-beta-D-glucopyranoside, 0,35% N-Decyl-beta-D-maltoside, 2% Lauryl-sarcosine, 0, 1% SDS, 0,3% CHAPS, 1M Guanidine HCl, 1mM PMSF, 10% Ethanol, 5M NaCl, 2M (NH4)SO₄, 1M CaCl₂, 25% Glycerol
  • Short-term storage: In equilibration buffer
  • Long-term storage: In 20% ethanol at 4 °C

The PureCube StrepTactin® MagBeads XL enable protein yields of up to 7 mg protein per ml MagBeads. They were developed for protein purification assays using the Strep-tag® using magnetic separation. PureCube StrepTactin® products can also be obtained as agarose resin.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty