Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HighSpec HighSpec Rho1D4 Agarose XL

Supplier: Cube Biotech
HighSpec
57602 57604 57603 57601
77393-338EA 448 USD InStock
77393-338 77393-340 77393-344 77393-342
HighSpec HighSpec Rho1D4 Agarose XL
Protein Purification Protein Purification Reagents
HighSpec Rho1D4 agarose XL resin is a suspension of 400 µm agarose beads coupled to an anti-Rho1D4 antibody.

  • Usage: Coupling biomolecules to active carboxy groups
  • Specificity: Exposed amine (NH₂) groups on biomolecule
  • Amine group density: Higher than 15 µmol/ml
  • Bead ligand: Chemically active Carboxy groups
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • Matrix: 6% cross-linked agarose
  • Short term storage: In equilibration buffer (see protocol)
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - Three years upon delivery, unopened
  • Chemical structure: Carboxy structure

The considerable size of the beads is meant for very viscous cell media and high flow rates. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. Additionally, we offer Rho1D4-tag affinity resin with a bead diameter of 40 µm for slightly higher protein yield and more mechanical stability of the beads. Alternatively, Rho1D4 MagBeads XL are available if you prefer this affinity chromatography method. Additional support products for Rho1D4-tag purification assays are Rho1D4 peptide, an essential compound of the elution buffer, and our Rho1D4 antibody for later western blot detection of your protein.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty