Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HighSpec Rho1D4 MagBeads XL

Supplier: Cube Biotech
HighSpec
57705 57701 57725
77393-350EA 6596 USD InStock
77393-350 77393-348 77393-346
HighSpec Rho1D4 MagBeads XL
Protein Purification Protein Purification Magnetic Beads
HighSpec Rho1D4 MagBeads XL is a suspension of 90 µm magnetic agarose beads coupled to an anti-Rho1D4 antibody.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead Ligand: anti-Rho1D4 antibody
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C

HighSpec Rho1D4 MagBeads XL resin is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. Additionally, we offer Rho1D4-tag affinity resin with a bead diameter of 30 µm for more mechanical stability and slightly higher protein yield. Alternatively, Rho1D4 Agarose resin is also available if you prefer this affinity chromatography method. Additional support products for Rho1D4-tag purification assays are Rho1D4 peptide, an essential compound of the elution buffer, and our Rho1D4 antibody for later western blot detection of your protein.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty