Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Carboxy MagBeads

Supplier: Cube Biotech
PureCube
58205 58290 58201 58225
77393-362EA 768 USD InStock
77393-362 77393-360 77393-366 77393-364
PureCube Carboxy MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube amine carboxy agarose MagBeads are your best option to couple biomolecules that carry free-standing Amine (NH₂) groups.

  • Usage: Coupling proteins to active Epoxy groups
  • Specificity: Exposed amine (NH₂) or thiol (SH) groups on biomolecule
  • Bead ligand: Chemically active Epoxy groups
  • Epoxy group density: 20 µmol/ml
  • Bead size: 400 μm
  • Filling quantity: Delivered as a 50% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In equilibration buffer (see protocol) at 4 °C
  • Long term storage: Best lyophilized, or in 100% isopropanol at 4 °C
  • Shelf life: If properly stored - six months upon delivery, unopened
  • Chemical structure: Epoxy structure

The resulting peptide bond between the Carboxy MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube carboxy MagBeads are also available as agarose resin in case you prefer this purification method or as normal-sized MagBeads if you would like to have more surface area on your beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty