Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube NHS Activated MagBeads XL

Supplier: Cube Biotech
PureCube
58490 58425 58405
77393-380PK 11946 USD InStock
77393-380 77393-376 77393-378
PureCube NHS Activated MagBeads XL
Protein Purification Protein Purification Magnetic Beads
PureCube NHS (N-hydroxy succinimide) MagBeads/magnetic beads XL are your best option to couple biomolecules carrying free-standing amine groups.

  • Usage: Coupling proteins to active NHS Groups
  • Specificity: Exposed amine (NH₂) groups on biomolecule
  • Binding capacity: >25 mg/ml
  • Bead ligand: Chemically active NHS groups
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 6% cross-linked agarose
  • Storage temperature: 4 °C
  • Shelf life: If properly stored - six months upon delivery, unopened
  • Chemical structure: NHS structure
  • Special notes: This product is shipped as a flammable good, 2-propanol UN1219

The resulting Amide bond between the NHS MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube NHS Activated MagBeads/magnetic beads are also available as agarose resin for FPLC or as MagBeads with a smaller bead size for more mechanical stability.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty