Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Epoxy Activated MagBeads XL

Supplier: Cube Biotech
PureCube
58805 58825 58890
77393-390EA 883 USD InStock
77393-390 77393-394 77393-392
PureCube Epoxy Activated MagBeads XL
Protein Purification Protein Purification Magnetic Beads
PureCube Epoxy Activated MagBeads XL is your best option to couple biomolecules that carry free-standing amine (NH₂) and thiol (SH) groups.

  • Usage: Coupling biomolecules to active Epoxy Groups
  • Specificity: Exposed amine (NH₂) or thiol (SH) groups on biomolecule
  • Bead ligand: Chemically active Epoxy groups
  • Epoxy group density: 20 µmol/ml
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 6% cross-linked agarose
  • Short term storage: In equilibration buffer (see protocol) at 4 °C
  • Long term storage: In 100% isopropanol at 4 °C
  • Shelf life: If properly stored - six months upon delivery, unopened
  • Chemical structure: Epoxy structure

The resulting amide and thiol bond between the Epoxy MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Epoxy Activated MagBeads XL are also available as agarose resin in case you prefer FPLC or batch spin methods or as MagBeads with a smaller bead diameter if your downstream application requires more mechanically stable beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty