Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Maleimide Activated Agarose XL

Supplier: Cube Biotech
PureCube
59103 59110 59112 59105
77393-408EA 635 USD InStock
77393-408 77393-410 77393-414 77393-412
PureCube Maleimide Activated Agarose XL
Protein Purification Protein Purification Reagents
PureCube maleimide agarose XL resin is your best option to couple biomolecules carrying thiol (SH) groups.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: 20 to 80 mg/ml, depending on bead size
  • Bead Ligand: Ni-NTA
  • Bead size: 40 μm, 100 µm, or 400 µm
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Filling quantity: Delivered as a 50% suspension

Required equipment:
- Lysis buffer
- Wash buffer
- Elution buffer
- Ice bath
- Refrigerated centrifuge for 50 ml tube (min 10000×g)
- 50 ml centrifuge tube
- Micropipettor and micropipetting tips
- Disposable gravity flow columns with capped bottom outlet, 2 ml
- pH meter
- End-over-end shaker
- SDS-PAGE buffers, reagents, and equipment optional: Western Blot reagents and equipment

The resulting Thioether bond between the maleimide agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube maleimide activated agarose is also available as magnetic beads in case you prefer this method, or as agarose resin with a smaller bead diameter if your downstream application requires more mechanically stable beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty