Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Maleimide Activated XL MagBeads

Supplier: Cube Biotech
PureCube
59290 59205 59225
77393-418EA 4355 USD InStock
77393-418 77393-416 77393-420
PureCube Maleimide Activated XL MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube maleimide MagBeads XL are your best option to couple biomolecules carrying thiol (SH) groups.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >80 mg/ml
  • Bead ligand: Ni-NTA
  • Bead size: 100 μm
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Filling quantity: Delivered as a 50% suspension

Required equipment and materials:
- FPLC hardware like e.g. ÄKTA
- Binding buffer
- Elution buffer

The resulting Thioether bond between the maleimide MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube maleimide activated MagBeads are also available as agarose resin in case you prefer this method catalog, or as MagBeads with a smaller bead diameter if your downstream application requires more mechanical stable beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty