Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube 100 Compact Cartridge Ni-NTA

Supplier: Cube Biotech
PureCube
74302 74306 74304 74308
77393-448EA 263 USD InStock
77393-448 77393-450 77393-446 77393-444
PureCube 100 Compact Cartridge Ni-NTA
Protein Purification Protein Purification Reagents
The PureCube 100 Ni-NTA compact cartridge is an IMAC chromatography column prepacked with PureCube 100 Ni-NTA resin.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead ligand: Co-NTA
  • Bead size: 100 μm
  • Filling quantity: Delivered as a 50% suspension

Required equipment and materials:
- FPLC hardware like e.g. ÄKTA
- Binding buffer
- Elution buffer

You can choose between two sizes, 1 ml bed volume and 5 ml bed volume. Please look at the given features table to ensure that the cartridge/column is compatible with your FPLC hardware like ÄKTA. If a compatibility cannot be ensured we recommend buying the sole loose PureCube 100 Ni-NTA resin and packing the columns yourself.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty