Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube INDIGO Ni-MagBeads

Supplier: Cube Biotech
PureCube
75290 75225
77393-502PK 5976 USD InStock
77393-502 77393-500
PureCube INDIGO Ni-MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Ni-INDIGO magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity: >100 mg/ml
  • Bead ligand: Ni-INDIGO
  • Bead size: 100 μm
  • Filling quantity: Delivered as a 50% suspension
  • pH stability: 2 to 14
  • Chelator stability (e.g. EDTA): 20 mM
  • Reducing agent stability (e.g. DTT): 20 mM
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Feature - PureCube mini batch spin column:
Body material: Polypropylene
Filter pore size: 0.1 to 0.2 µm low binding PVDF
Max. volume: 600 µl
Max. g force: 12000 to 14000×g (45-degree fixed angle rotor)
Min. g force: 2500×g for 1 min

Developed with the aim in mind of creating magnetic beads of the highest quality that can tolerate the highest concentrations of chelators like EDTA and DTT, which are often used in mammalian cell culture buffers. The resulting INDIGO ligand has EDTA and DTT stabilities of 20 mM each. That is highly superior to other competing magnetic beads like Ni-NTA.

In biotechnological and pharmaceutical processes in general, the INDIGO-Ni ligand has the advantage over the NTA ligand that no nickel leakage occurs due to its strong binding.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty