Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube 100 Compact Cartridge Ni-INDIGO

Supplier: Cube Biotech
75302 75304 75306 75308
77393-506PK 337 USD InStock
77393-506 77393-504 77393-508 77393-510
PureCube 100 Compact Cartridge Ni-INDIGO
Protein Purification Protein Purification Reagents
The PureCube Ni-INDIGO column/cartridge is an IMAC chromatography column prepacked with PureCube 100 INDIGO-Ni Agarose resin beads.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity: >100 mg/ml
  • Bead Ligand: Ni-INDIGO
  • Bead size: 100 μm
  • Filling quantity: Delivered as a 50% suspension
  • pH stability: 2 to 14
  • Chelator stability (e.g. EDTA): 20 mM
  • Reducing agent stability (e.g. DTT): 20 mM
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Features - PureCube mini batch spin column:
Body material: Polypropylene
Filter pore size: 0.1 to 0.2 µm low binding PVDF
Max. volume: 20 ml
Max. g force: 1500 to 2000×g (swing-bucket rotor)
Min. g force: 100 to 200×g for 1 to 10 min

You can choose between two sizes, 1 ml bed volume and 5 ml bed volume. The PureCube cartridge/column that is used here is compatible with almost all commonly used FPLC systems like ÄKTA. We further recommend checking out our other Ni-INDIGO products.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty