Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Cu-NTA Agarose

Supplier: Cube Biotech
PureCube
31403-CU 31401-CU 31412-CU 31410-CU 31405-CU
77393-528EA 174 USD InStock
77393-528 77393-536 77393-552 77393-520 77393-544
PureCube Cu-NTA Agarose
Protein Purification Protein Purification Reagents
Our PureCube Cu-NTA agarose resins are small agarose resin beads with a diameter of 40 µm.

  • Usage: Specific binding and purification of 6× His-tagged proteins, Binding of Copper-binding proteins
  • Specificity: Affinity to His-tagged proteins, affinity to Copper-binding proteins
  • Binding capacity: >80 mg/ml
  • Bead Ligand: Cu-NTA
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • Metal ion capacity: >15 µeqv Cu²⁺/ml
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
  • Required equipment:
  • Lysis Buffer
  • Wash Buffer
  • Elution Buffer
  • Ice bath
  • Refrigerated centrifuge for 50 ml tube (min 10000×g)
  • 50 ml centrifuge tube
  • Micropipettor and Micropipetting tips
  • Disposable gravity flow columns with capped bottom outlet, 2 ml
  • pH meter
  • End-over-end shaker
  • SDS-PAGE buffers, reagents and equipment Optional: Western Blot reagents and equipment

PureCube Cu-NTA agarose resins are used for the purification of active His-tagged proteins from cells and the enrichment of Copper-binding proteins. Our year-long experience in manufacturing agarose resin led to the high yield of 80 mg protein per ml resin, which is leading in the market compared to other Cu-NTA suppliers. PureCube Cu-NTA resins are suited for batch spin columns and FPLC. Their small diameter provides them with great mechanical stability.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty