Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Al-NTA Agarose

Supplier: Cube Biotech
PureCube
31412-AL 31403-AL 31410-AL 31401-AL 31405-AL
77393-526EA 174 USD InStock
77393-526 77393-518 77393-542 77393-550 77393-534
PureCube Al-NTA Agarose
Protein Purification Protein Purification Reagents
PureCube Al-NTA agarose resin beads were developed for the enrichment of phosphorylated proteins.

  • Usage: Enrichment of phosphopeptides
  • Specificity: Affinity to phosphorylated biomolecules
  • Bead ligand: Al-NTA
  • Bead size: 40 µm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% crosslinked agarose
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

PureCube Al-NTA agarose resin beads can also be used for the purification of His-tagged proteins as a secondary function. The affinity matrix is based on 7.5% cross-linked agarose that was magnetized. The bead ligand is Al-NTA and the particle diameter is 40 µm. They are very homogeneous in size, yielding a high degree of reproducibility.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty