Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Cu-NTA MagBeads

Supplier: Cube Biotech
PureCube
31590-CU 31501-CU 31591-CU 31505-CU 31525-CU
77393-608EA 5528 USD InStock
77393-608 77393-560 77393-596 77393-572 77393-584
PureCube Cu-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Cu-NTA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.

  • Usage: Specific binding and purification of 6× His-tagged proteins, binding of copper-binding proteins
  • Specificity: Affinity to His-tagged proteins, Affinity to Copper-finger proteins
  • Binding capacity: >80 mg/ml
  • Bead ligand: Cu-NTA
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Metal ion capacity: >15 µeqv Zn²⁺/ml
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Our year-long experience in manufacturing agarose resin lead to the high yield of 50 mg protein per ml resin, which is leading in the market compared to other Cu-NTA suppliers.

Cu-NTA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin. For your convenience Cube Biotech also offers its own MagBead separator for use with our magnetic beads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty