Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Ti-NTA MagBeads

Supplier: Cube Biotech
PureCube
31505-TI 31525-TI 31590-TI
77393-600PK 5719 USD InStock
77393-600 77393-576 77393-588
PureCube Ti-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Ti-NTA MagBeads/magnetic beads were developed for the enrichment of phosphorylated proteins. They can also be used for the purification of His-tagged proteins as a secondary function.

  • Usage: Specific binding and purification of 6× His-tagged proteins, binding of copper-binding proteins
  • Specificity: Affinity to His-tagged proteins, affinity to copper-binding
  • Binding capacity: >80 mg/ml
  • Bead Ligand: Cu-NTA
  • Bead size: 40 μm
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Filling quantity: Delivered as a 50% suspension

Required equipment and materials:
- FPLC hardware like e.g. AKT
- Binding buffer
- Elution buffer

The affinity matrix is based on 6% cross-linked agarose that was magnetized. The bead ligand is Ti-NTA and the particle diameter is 30 µm. They are very homogeneous in size, yielding in a high degree of reproducibility. The phosphopeptide enrichment with Cube Biotech MagBeads can be automated.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty