Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Zr-NTA MagBeads

Supplier: Cube Biotech
PureCube
31590-ZR 31501-ZR 31505-ZR 31525-ZR
77393-604PK 5719 USD InStock
77393-604 77393-592 77393-580 77393-568
PureCube Zr-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads
PureCube Zr-NTA MagBeads/magnetic beads were developed for the enrichment of phosphorylated proteins. They can also be used for the purification of His-tagged proteins as a secondary function.

  • Usage: Enrichment of phosphopeptides, specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Affinity to phosphorylated biomolecules
  • Bead ligand: Fe-NTA
  • Bead size: 40 µm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% crosslinked agarose
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

The affinity matrix is based on 6% cross-linked agarose that was magnetized. The bead ligand is Zr-NTA and the particle diameter is 30 µm. They are very homogeneous in size, yielding in a high degree of reproducibility. The phosphopeptide enrichment with Cube Biotech MagBeads can be automated as it has already been proven in this paper.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty