Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Al-NTA

Supplier: Cube Biotech
PureCube
31604-AL 31608-AL 31602-AL 31606-AL
77393-638PK 886 USD InStock
77393-638 77393-614 77393-630 77393-622
PureCube Compact Cartridge Al-NTA
Protein Purification Protein Purification Reagents
PureCube Al-NTA compact cartridge is an FPLC-ready column pre-packed with our PureCube Al-NTA agarose resin. It is used for the purification of His-tagged proteins and the enrichment of phosphorylated proteins.

  • Usage:Enrichment of phosphopeptides, specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to phosphorylated biomolecules
  • Bead ligand: Al-NTA
  • Bead size: 40 µm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% crosslinked agarose
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

As an alternative, we also offer PureCube Al-NTA MagBeads, the more common format of Fe-NTA beads for phosphopeptide enrichment.

The PureCube cartridge/column that is used here is compatible with almost all commonly used FPLC systems like ÄKTA TM.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty