Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Cu-NTA

Supplier: Cube Biotech
PureCube
31606-CU 31602-CU 31604-CU 31608-CU
77393-616EA 62 USD InStock
77393-616 77393-624 77393-640 77393-632
PureCube Compact Cartridge Cu-NTA
Protein Purification Protein Purification Reagents
The PureCube Cu-NTA compact cartridge is an IMAC chromatography column prepacked with PureCube Cu-NTA resin. You can choose between two sizes, 1 ml bed volume and 5 ml bed volume.

  • Purity: >90% (determined by SDS-PAGE)
  • Number of amino acids: 217
  • Molecular mass: 25309 Da
  • Extinction coefficient (in water) ε280: 25440 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (Human origin): MGSSHHHHHH ENLYFQ GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
  • Affinity tag: N-Terminal His-tag

Please have a look at the given features to ensure that the cartridge/column is compatible with your FPLC hardware like ÄKTA TM. If this is not the case we recommend buying the sole loose PureCube Cu-NTA resin and packing the columns yourself.

Alternatively, we also offer our PureCube Cu-NTA products as magnetic beads, if you prefer this IMAC method.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty