Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Compact Cartridge Zn-NTA

Supplier: Cube Biotech
PureCube
31604-ZN 31606-ZN 31608-ZN 31602-ZN
77393-636EA 187 USD InStock
77393-636 77393-628 77393-644 77393-620
PureCube Compact Cartridge Zn-NTA
Protein Purification Protein Purification Reagents
The PureCube Zn-NTA Compact cartridge is an IMAC chromatography column prepacked with PureCube Zn-NTA resin. You can choose between two sizes, 1 and 5 ml bed volume.

  • Usage: Specific binding and purification of 6× His-tagged proteins
  • Binding of Zinc-binding proteins
  • Specificity: Affinity to His-tagged proteins
  • Affinity to Zinc-finger proteins
  • Binding capacity: >50 mg/ml
  • Bead Ligand: Zn-NTA
  • Bead size: 40 μm
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • Filling quantity: Delivered as a 50% suspension
  • Required equipment and materials:
  • FPLC hardware like e.g. ÄKTA
  • Binding Buffer
  • Elution Buffer

Please have a look at the given features table to ensure that the Cartridge/Column is compatible with your FPLC hardware. Common systems like ÄKTA TM are compatible without problem. If this is not the case we recommend buying the sole loose PureCube Zn-NTA resin and packing the columns yourself. Alternatively, we also offer our PureCube Zn-NTA products as magnetic beads, if you prefer this IMAC method.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty