Would you like to log in again?
Sterilizable Goggles, 611 Polycarbonate AF121
Supplier: Univet
611.S0.00.01 611.S1.00.01
77393-712CS 4300 USD InStock
77393-712 77393-718
Sterilizable Goggles, 611 Polycarbonate AF121
Goggles
611 Polycarbonate AF121 sterilizable goggles are available in direct vent or indirect vent formats. Both formats feature an anti-fog and anti-scratch lens treatment.
  • Goggles feature interchangeable lens technology
  • Meets EN 166 and EN 170 standards
  • Scale number: (2C-1.2)
  • Mechanical resistance: Medium energy impact (B)
  • Optical class (1)
Goggles with polycarbonate lens are reusable for up to 40 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. The AF121 treatment applied to the lens prevents scratches and fogging at the same time. Goggles will withstand up to 10 steam sterilization cycles. They are also reusable for up to 10 gamma irradiation cycles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty