Would you like to log in again?
Sterilizable Overspec Goggles, 619 Polycarbonate
Supplier: Univet
619.04.23.10 619.05.23.10
77393-724CS 4000 USD InStock
77393-724 77393-722
Sterilizable Overspec Goggles, 619 Polycarbonate
Goggles
619 Polycarbonate sterilizable overspec goggles are available in direct vent or indirect vent formats.
  • Overspec goggles feature interchangeable lens technology
  • Meets EN 166 and EN 170 standards
  • Scale number: (2C-1.2)
  • Mechanical resistance: Medium energy impact (B)
  • Optical class (1)
Goggles with polycarbonate lens are reusable for up to 30 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. They are also reusable for up to 10 gamma irradiation cycles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty